Search
Search

ATXN7L1| Gene Symbol: ATXN7L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012176688.2 | XP_012032078.2 | 7 - 8 | 1230 | 64 | |
| XM_012176690.2 | XP_012032080.2 | 7 - 8 | 1230 | 64 | ||
| XM_012176693.2 | XP_012032083.2 | 7 - 8 | 1230 | 64 | ||
| XM_012176694.2 | XP_012032084.2 | 7 - 8 | 1230 | 64 | ||
| XM_012176695.2 | XP_012032085.2 | 5 - 6 | 1318 | 64 | ||
Set Membership: |
|