Search
Search

ZNF202| Gene Symbol: ZNF202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016117.4 | XP_004016166.2 | 4 - 5 | 904 | 57 | |
| XM_012095651.3 | XP_011951041.2 | 4 - 5 | 831 | 57 | ||
| XM_015100973.2 | XP_014956459.2 | 4 - 5 | 820 | 57 | ||
| XM_027979345.1 | XP_027835146.1 | 2 - 3 | 149 | 57 | ||
| XM_027979346.1 | XP_027835147.1 | 2 - 3 | 157 | 57 | ||
Set Membership: |
|