Search
Search

SLC8B1| Gene Symbol: SLC8B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956571.1 | XP_027812372.1 | 6 - 7 | 1007 | 56 | |
| XM_027956572.1 | XP_027812373.1 | 6 - 7 | 1004 | 56 | ||
Set Membership: |
|