Search
Search

SLC25A12| Gene Symbol: SLC25A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004597.4 | XP_004004646.1 | 6 - 7 | 688 | 61 | |
| XM_012133795.3 | XP_011989185.1 | 6 - 7 | 719 | 61 | ||
Set Membership: |
|