Search
Search

CSF3R| Gene Symbol: CSF3R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967959.1 | XP_027823760.1 | 14 - 15 | 2037 | 59 | |
| XM_027967969.1 | XP_027823770.1 | 14 - 15 | 2037 | 59 | ||
| XM_027967974.1 | XP_027823775.1 | 14 - 15 | 2037 | 59 | ||
| XM_027967981.1 | XP_027823782.1 | 14 - 15 | 2037 | 59 | ||
Set Membership: |
|