Search
Search

MED18| Gene Symbol: MED18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005069.4 | XP_004005118.1 | 2 - 3 | 350 | 102 | |
| XM_012150747.3 | XP_012006137.1 | 2 - 3 | 319 | 102 | ||
| XM_015093651.2 | XP_014949137.1 | 2 - 3 | 244 | 102 | ||
Set Membership: |
|