Search
Search

TAF1A| Gene Symbol: TAF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013612.4 | XP_004013661.2 | 5 - 6 | 737 | 101 | |
| XM_012187080.3 | XP_012042470.2 | 5 - 6 | 703 | 101 | ||
Set Membership: |
|