Search
Search
| Gene Symbol: ETV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012176421.3 | XP_012031811.1 | 6 - 7 | 749 | 76 | |
| XM_012176422.3 | XP_012031812.1 | 8 - 9 | 1312 | 76 | ||
| XM_012176423.3 | XP_012031813.1 | 7 - 8 | 1115 | 76 | ||
| XM_012176426.3 | XP_012031816.1 | 5 - 6 | 1469 | 76 | ||
Set Membership: |
|