Search
Search

GCFC2| Gene Symbol: GCFC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005926.4 | XP_004005975.2 | 15 - 16 | 2110 | 77 | |
| XM_012170683.3 | XP_012026073.2 | 15 - 16 | 2086 | 77 | ||
Set Membership: |
|