Search
Search

TRIM2| Gene Symbol: TRIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017189.4 | XP_004017238.3 | 2 - 3 | 348 | 56 | |
| XM_012097332.3 | XP_011952722.2 | 2 - 3 | 391 | 56 | ||
| XM_012097333.3 | XP_011952723.2 | 2 - 3 | 427 | 56 | ||
| XM_012097334.3 | XP_011952724.2 | 2 - 3 | 424 | 56 | ||
| XM_027956229.1 | XP_027812030.1 | 2 - 3 | 316 | 56 | ||
| XM_027956230.1 | XP_027812031.1 | 2 - 3 | 313 | 56 | ||
| XM_027956231.1 | XP_027812032.1 | 2 - 3 | 428 | 56 | ||
| XM_027956232.1 | XP_027812033.1 | 2 - 3 | 425 | 56 | ||
Set Membership: |
|