Search
Search

ABCC11| Gene Symbol: ABCC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977814.1 | XP_027833615.1 | 28 - 29 | 4047 | 62 | |
| XM_027977815.1 | XP_027833616.1 | 27 - 28 | 3933 | 62 | ||
Set Membership: |
|