Search
Search

C18H14orf28| Gene Symbol: C18H14orf28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102016.2 | XP_014957502.1 | 2 - 3 | 803 | 71 | |
| XM_015102018.2 | XP_014957504.1 | 2 - 3 | 603 | 71 | ||
| XM_027957298.1 | XP_027813099.1 | 2 - 3 | 605 | 71 | ||
Set Membership: |
|