Search
Search

MYH10| Gene Symbol: MYH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974902.1 | XP_027830703.1 | 22 - 23 | 2892 | 53 | |
| XM_027974903.1 | XP_027830704.1 | 22 - 23 | 2669 | 53 | ||
| XM_027974904.1 | XP_027830705.1 | 21 - 22 | 2861 | 53 | ||
| XM_027974905.1 | XP_027830706.1 | 21 - 22 | 2828 | 53 | ||
| XM_027974906.1 | XP_027830707.1 | 20 - 21 | 2798 | 53 | ||
Set Membership: |
|