Search
Search

RAPGEF5| Gene Symbol: RAPGEF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968507.1 | XP_027824308.1 | 15 - 16 | 1493 | 63 | |
| XM_027968508.1 | XP_027824309.1 | 10 - 11 | 1342 | 63 | ||
Set Membership: |
|