Search
Search

ARMH3| Gene Symbol: ARMH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020136.4 | XP_004020185.1 | 12 - 13 | 1014 | 83 | |
| XM_012102828.3 | XP_011958218.1 | 13 - 14 | 1040 | 83 | ||
| XM_027960466.1 | XP_027816267.1 | 13 - 14 | 1112 | 83 | ||