Search
Search

TPD52| Gene Symbol: TPD52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011758.4 | XP_004011807.1 | 3 - 4 | 412 | 72 | |
| XM_012183906.3 | XP_012039296.1 | 3 - 4 | 429 | 72 | ||
| XM_012183907.3 | XP_012039297.1 | 3 - 4 | 426 | 72 | ||
| XM_027973078.1 | XP_027828879.1 | 3 - 4 | 331 | 72 | ||
| XM_027973079.1 | XP_027828880.1 | 3 - 4 | 331 | 72 | ||
| XM_027973080.1 | XP_027828881.1 | 3 - 4 | 436 | 72 | ||
Set Membership: |
|