Search
Search

DIAPH2| Gene Symbol: DIAPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012107728.3 | XP_011963118.3 | 23 - 24 | 2966 | 68 | |
| XM_015104666.2 | XP_014960152.2 | 22 - 23 | 2945 | 68 | ||
| XM_015104667.2 | XP_014960153.2 | 23 - 24 | 2967 | 68 | ||
| XM_015104668.2 | XP_014960154.2 | 22 - 23 | 2933 | 68 | ||
| XM_015104669.2 | XP_014960155.2 | 20 - 21 | 2388 | 68 | ||
Set Membership: |
|