Search
Search

MAPK6| Gene Symbol: MAPK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012181550.3 | XP_012036940.1 | 4 - 5 | 1590 | 76 | |
| XM_027971825.1 | XP_027827626.1 | 4 - 5 | 1640 | 76 | ||
Set Membership: |
|