Search
Search

RPGRIP1L| Gene Symbol: RPGRIP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012189787.3 | XP_012045177.1 | 25 - 26 | 3771 | 67 | |
| XM_012189788.3 | XP_012045178.1 | 25 - 26 | 3753 | 67 | ||
| XM_012189789.3 | XP_012045179.1 | 24 - 25 | 3318 | 67 | ||
Set Membership: |
|