Search
Search

LRRC45| Gene Symbol: LRRC45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013057.4 | XP_004013106.2 | 2 - 3 | 709 | 51 | |
| XM_012186412.3 | XP_012041802.2 | 2 - 3 | 709 | 51 | ||
Set Membership: |
|