Search
Search

BCORL1| Gene Symbol: BCORL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963619.1 | XP_027819420.1 | 13 - 14 | 5125 | 61 | |
| XM_027963620.1 | XP_027819421.1 | 13 - 14 | 5127 | 61 | ||
| XM_027963621.1 | XP_027819422.1 | 12 - 13 | 5109 | 61 | ||
| XM_027963622.1 | XP_027819423.1 | 12 - 13 | 5046 | 61 | ||
| XM_027963623.1 | XP_027819424.1 | 12 - 13 | 5044 | 61 | ||
Set Membership: |
|