Search
Search

ANXA6| Gene Symbol: ANXA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008987.3 | XP_004009036.1 | 9 - 10 | 794 | 59 | |
| XM_027970423.1 | XP_027826224.1 | 8 - 9 | 639 | 59 | ||
| XM_027970424.1 | XP_027826225.1 | 8 - 9 | 643 | 59 | ||
Set Membership: |
|