Search
Search

CASP3
LOC114116869| Gene Symbol: CASP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104559.2 | XP_014960045.1 | 6 - 7 | 695 | 77 | |
| XM_027962551.1 | XP_027818352.1 | 6 - 7 | 708 | 77 | ||
| Gene Symbol: LOC114116869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974864.1 | XP_027830665.1 | 1 | 697 | 77 | |
Set Membership: |
|