Search
Search

TDRD10| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012182462.3 | XP_012037852.2 | 5 - 6 | 317 | 98 | |
| XM_015092270.2 | XP_014947756.2 | 4 - 5 | 1838 | 98 | ||
Set Membership: |
|