Search
Search

DAZAP1| Gene Symbol: DAZAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970193.1 | XP_027825994.1 | 10 - 11 | 1004 | 67 | |
| XM_027970194.1 | XP_027825995.1 | 10 - 11 | 1003 | 67 | ||
| XM_027970195.1 | XP_027825996.1 | 7 - 8 | 713 | 67 | ||
| XM_027970196.1 | XP_027825997.1 | 7 - 8 | 723 | 67 | ||
| XM_027970197.1 | XP_027825998.1 | 10 - 11 | 1008 | 67 | ||
Set Membership: |
|