Search
Search

C21H11orf80| Gene Symbol: C21H11orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959697.1 | XP_027815498.1 | 4 - 5 | 383 | 70 | |
| XM_027959698.1 | XP_027815499.1 | 4 - 5 | 383 | 70 | ||
| XM_027959699.1 | XP_027815500.1 | 4 - 5 | 305 | 70 | ||
Set Membership: |
|