Search
Search

FERMT3| Gene Symbol: FERMT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959374.1 | XP_027815175.1 | 11 - 12 | 1524 | 61 | |
| XM_027959375.1 | XP_027815176.1 | 11 - 12 | 1532 | 61 | ||
| XM_027959376.1 | XP_027815177.1 | 11 - 12 | 1478 | 61 | ||
Set Membership: |
|