Search
Search

COL5A1| Gene Symbol: COL5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966117.1 | XP_027821918.1 | 60 - 61 | 5084 | 55 | |
| XM_027966118.1 | XP_027821919.1 | 60 - 61 | 5085 | 55 | ||
Set Membership: |
|