Search
Search

GEMIN5| Gene Symbol: GEMIN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009008.4 | XP_004009057.2 | 20 - 21 | 2928 | 57 | |
| XM_012178843.3 | XP_012034233.2 | 20 - 21 | 2931 | 57 | ||
Set Membership: |
|