Search
Search
| Gene Symbol: STN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012102876.2 | XP_011958266.1 | 3 - 4 | 441 | 111 | |
| XM_012102877.3 | XP_011958267.1 | 3 - 4 | 328 | 111 | ||
| XM_012102878.2 | XP_011958268.1 | 3 - 4 | 457 | 111 | ||
| XM_012102879.2 | XP_011958269.1 | 3 - 4 | 441 | 111 | ||
| XM_012102881.2 | XP_011958271.1 | 3 - 4 | 441 | 111 | ||
| XM_027960169.1 | XP_027815970.1 | 3 - 4 | 328 | 111 | ||
Set Membership: |
|