Search
Search

APEX2| Gene Symbol: APEX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106626.3 | XP_011962016.1 | 3 - 4 | 316 | 75 | |
| XM_012106627.3 | XP_011962017.1 | 4 - 5 | 491 | 75 | ||
| XM_027963026.1 | XP_027818827.1 | 4 - 5 | 694 | 75 | ||