Search
Search

NBEAL1| Gene Symbol: NBEAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093496.2 | XP_014948982.2 | 42 - 43 | 6798 | 63 | |
| XM_015093498.2 | XP_014948984.2 | 42 - 43 | 6798 | 63 | ||
| XM_027965103.1 | XP_027820904.1 | 43 - 44 | 7224 | 63 | ||
| XM_027965104.1 | XP_027820905.1 | 43 - 44 | 6871 | 63 | ||
| XM_027965105.1 | XP_027820906.1 | 42 - 43 | 6861 | 63 | ||
| XM_027965106.1 | XP_027820907.1 | 41 - 42 | 6711 | 63 | ||
| XM_027965108.1 | XP_027820909.1 | 41 - 42 | 6711 | 63 | ||
Set Membership: |
|