Search
Search

CCDC78| Gene Symbol: CCDC78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961334.1 | XP_027817135.1 | 6 - 7 | 1066 | 75 | |
| XM_027961335.1 | XP_027817136.1 | 6 - 7 | 1068 | 75 | ||
| XM_027961336.1 | XP_027817137.1 | 6 - 7 | 1068 | 75 | ||
| XM_027961337.1 | XP_027817138.1 | 6 - 7 | 1069 | 75 | ||
| XM_027961338.1 | XP_027817139.1 | 6 - 7 | 1069 | 75 | ||
Set Membership: |
|