Search
Search
| Gene Symbol: RHOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973200.1 | XP_027829001.1 | 3 - 4 | 374 | 118 | |
| XM_027973203.1 | XP_027829004.1 | 3 - 4 | 840 | 118 | ||
| XM_027973209.1 | XP_027829010.1 | 3 - 4 | 919 | 118 | ||
Set Membership: |
|