Search
Search

CCDC33| Gene Symbol: CCDC33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101990.2 | XP_014957476.1 | 5 - 6 | 635 | 51 | |
| XM_027957196.1 | XP_027812997.1 | 5 - 6 | 1133 | 51 | ||
| XM_027957197.1 | XP_027812998.1 | 5 - 6 | 1137 | 51 | ||
| XM_027957198.1 | XP_027812999.1 | 5 - 6 | 1133 | 51 | ||
Set Membership: |
|