Search
Search
| Gene Symbol: AGER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018915.4 | XP_004018964.2 | 9 - 10 | 1434 | 55 | |
| XM_004018916.4 | XP_004018965.2 | 9 - 10 | 1467 | 55 | ||
| XM_012100908.3 | XP_011956298.2 | 9 - 10 | 1446 | 55 | ||
| XM_012100909.3 | XP_011956299.1 | 8 - 9 | 1349 | 55 | ||
| XM_012100910.2 | XP_011956300.1 | 7 - 8 | 903 | 55 | ||
Set Membership: |
|