Search
Search

NCSTN| Gene Symbol: NCSTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002671.3 | XP_004002720.2 | 12 - 13 | 1517 | 61 | |
| XM_012184363.3 | XP_012039753.2 | 12 - 13 | 1845 | 61 | ||
Set Membership: |
|