Search
Search

DLGAP5| Gene Symbol: DLGAP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010665.4 | XP_004010714.2 | 4 - 5 | 593 | 65 | |
| XM_012181687.3 | XP_012037077.2 | 4 - 5 | 593 | 65 | ||
Set Membership: |
|