Search
Search

PIK3CB| Gene Symbol: PIK3CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003302.4 | XP_004003351.2 | 15 - 16 | 2283 | 70 | |
| XM_012102527.3 | XP_011957917.2 | 13 - 14 | 2029 | 70 | ||
Set Membership: |
|