Search
Search

TSPAN19| Gene Symbol: TSPAN19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006232.4 | XP_004006281.2 | 4 - 5 | 341 | 77 | |
| XM_004006233.4 | XP_004006282.2 | 4 - 5 | 311 | 77 | ||
| XM_012174038.3 | XP_012029428.2 | 5 - 6 | 425 | 77 | ||
Set Membership: |
|