Search
Search
| Gene Symbol: ART3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180191.3 | XP_012035581.2 | 2 - 3 | 200 | 103 | |
| XM_012180193.3 | XP_012035583.2 | 2 - 3 | 181 | 103 | ||
| XM_012180194.3 | XP_012035584.2 | 2 - 3 | 200 | 103 | ||
| XM_012180196.3 | XP_012035586.2 | 2 - 3 | 200 | 103 | ||
| XM_012180197.3 | XP_012035587.2 | 2 - 3 | 200 | 103 | ||
| XM_012180198.3 | XP_012035588.2 | 2 - 3 | 200 | 103 | ||
| XM_012180201.2 | XP_012035591.2 | 2 - 3 | 202 | 103 | ||
| XM_027971096.1 | XP_027826897.1 | 2 - 3 | 200 | 103 | ||
| XM_027971097.1 | XP_027826898.1 | 2 - 3 | 201 | 103 | ||
| XM_027971098.1 | XP_027826899.1 | 2 - 3 | 200 | 103 | ||
| XM_027971099.1 | XP_027826900.1 | 2 - 3 | 201 | 103 | ||
| XM_027971100.1 | XP_027826901.1 | 2 - 3 | 201 | 103 | ||
| XM_027971101.1 | XP_027826902.1 | 2 - 3 | 200 | 103 | ||
| XM_027971102.1 | XP_027826903.1 | 2 - 3 | 202 | 103 | ||
| XM_027971103.1 | XP_027826904.1 | 2 - 3 | 200 | 103 | ||
Set Membership: |
|