Search
Search
| Gene Symbol: ARSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973987.1 | XP_027829788.1 | 6 - 7 | 1261 | 86 | |
| XM_027973988.1 | XP_027829789.1 | 6 - 7 | 1258 | 86 | ||
| XM_027973989.1 | XP_027829790.1 | 6 - 7 | 702 | 86 | ||
| XM_027973990.1 | XP_027829791.1 | 6 - 7 | 1256 | 86 | ||
| XM_027973991.1 | XP_027829792.1 | 3 - 4 | 916 | 86 | ||
| XM_027973992.1 | XP_027829793.1 | 6 - 7 | 1253 | 86 | ||
Set Membership: |
|