Search
Search

LOC101113264| Gene Symbol: LOC101113264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977792.1 | XP_027833593.1 | 6 - 7 | 709 | 88 | |
| XM_027977793.1 | XP_027833594.1 | 6 - 7 | 675 | 88 | ||
Set Membership: |
|