Search
Search

LOC101111337| Gene Symbol: LOC101111337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012183710.3 | XP_012039100.2 | 4 - 5 | 988 | 85 | |
| XM_012183728.3 | XP_012039118.1 | 3 - 4 | 874 | 85 | ||
Set Membership: |
|