Search
Search

MPV17L| Gene Symbol: MPV17L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020788.4 | XP_004020837.1 | 2 - 3 | 441 | 116 | |
| XM_027961630.1 | XP_027817431.1 | 2 - 3 | 383 | 116 | ||
Set Membership: |
|