Search
Search

FRMD4B| Gene Symbol: FRMD4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958177.1 | XP_027813978.1 | 20 - 21 | 2089 | 59 | |
| XM_027958178.1 | XP_027813979.1 | 20 - 21 | 2161 | 59 | ||
Set Membership: |
|