Search
Search

CCDC92| Gene Symbol: CCDC92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101581.2 | XP_014957067.1 | 3 - 4 | 537 | 113 | |
| XM_027956419.1 | XP_027812220.1 | 3 - 4 | 2888 | 113 | ||
| XM_027956420.1 | XP_027812221.1 | 3 - 4 | 254 | 113 | ||
| XM_027956421.1 | XP_027812222.1 | 4 - 5 | 276 | 113 | ||
Set Membership: |
|