Search
Search
| Gene Symbol: NSMF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966038.1 | XP_027821839.1 | 9 - 10 | 1301 | 64 | |
| XM_027966039.1 | XP_027821840.1 | 9 - 10 | 1277 | 64 | ||
| XM_027966040.1 | XP_027821841.1 | 9 - 10 | 1238 | 64 | ||
| XM_027966041.1 | XP_027821842.1 | 8 - 9 | 1207 | 64 | ||
| XM_027966042.1 | XP_027821843.1 | 9 - 10 | 1214 | 64 | ||
| XM_027966043.1 | XP_027821844.1 | 8 - 9 | 1180 | 64 | ||
Set Membership: |
|