Search
Search

LOC101102009
LOC114118322| Gene Symbol: LOC101102009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016472.4 | XP_004016521.1 | 1 | 480 | 85 | |
| Gene Symbol: LOC114118322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979070.1 | XP_027834871.1 | 1 | 477 | 85 | |
Set Membership: |
|