Search
Search

TRPC6| Gene Symbol: TRPC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708603.2 | XP_002708649.1 | 4 - 5 | 1783 | 80 | |
| XM_008262243.1 | XP_008260465.1 | 4 - 5 | 1783 | 80 | ||
Set Membership: |
|